Assay Details
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315607.1 | NP_001302536.1 | 10 - 11 | 1489 | 110 | |
XM_005655659.3 | XP_005655716.1 | 1617 | 110 | |||
XM_021091087.1 | XP_020946746.1 | 1651 | 110 | |||
XM_021091089.1 | XP_020946748.1 | 1783 | 110 | |||
XM_021091090.1 | XP_020946749.1 | 1788 | 110 | |||
XM_021091091.1 | XP_020946750.1 | 1473 | 110 | |||
GenBank mRNA | LN833582.1 | - | 10 - 11 | 1489 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
