Assay Details
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315607.1 | NP_001302536.1 | 5 - 6 | 628 | 96 | |
XM_005655659.3 | XP_005655716.1 | 756 | 96 | |||
XM_021091087.1 | XP_020946746.1 | 790 | 96 | |||
XM_021091089.1 | XP_020946748.1 | 922 | 96 | |||
XM_021091090.1 | XP_020946749.1 | 927 | 96 | |||
XM_021091091.1 | XP_020946750.1 | 612 | 96 | |||
GenBank mRNA | AK394342.1 | - | 3 - 4 | 397 | 96 | |
LN833582.1 | - | 5 - 6 | 628 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
