Assay Details
Gene Symbol: TENT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315609.1 | NP_001302538.1 | 1 - 2 | 109 | 116 | |
GenBank mRNA | AK348379.1 | - | 1 - 2 | 102 | 116 | |
AK348553.1 | - | 1 - 2 | 109 | 116 | ||
KJ820676.1 | - | 1 - 2 | 109 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001315609.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
