Assay Details
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315813.1 | NP_001302742.1 | 3 - 4 | 382 | 106 | |
XM_021085710.1 | XP_020941369.1 | 636 | 106 | |||
XM_021085711.1 | XP_020941370.1 | 631 | 106 | |||
XM_021085712.1 | XP_020941371.1 | 654 | 106 | |||
GenBank mRNA | JF944894.1 | - | 3 - 4 | 382 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021085712.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
