Assay Details
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087989.1 | XP_020943648.1 | 1310 | 74 | ||
XM_021087990.1 | XP_020943649.1 | 1304 | 74 | |||
XM_021087991.1 | XP_020943650.1 | 1028 | 74 | |||
XM_021087992.1 | XP_020943651.1 | 675 | 74 | |||
XM_021087994.1 | XP_020943653.1 | 733 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
