Assay Details
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482448.4 | XP_003482496.1 | 575 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003482448.4 transcripts.
Set Membership: |
![]() ![]() |
