Assay Details
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005657807.3 | XP_005657864.1 | 265 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
