Assay Details
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013983489.2 | XP_013838943.2 | 523 | 108 | ||
XM_021072811.1 | XP_020928470.1 | 496 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021072811.1 transcripts.
Set Membership: |
![]() ![]() |
