Assay Details
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013985912.2 | XP_013841366.2 | 1434 | 76 | ||
XM_021080515.1 | XP_020936174.1 | 1437 | 76 | |||
XM_021080516.1 | XP_020936175.1 | 1606 | 76 | |||
XM_021080517.1 | XP_020936176.1 | 745 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
