Assay Details
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013994960.2 | XP_013850414.2 | 251 | 107 | ||
XM_021094517.1 | XP_020950176.1 | 728 | 107 |

More Information
Set Membership: |
![]() ![]() |
