Assay Details
Gene Symbol: THEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013997133.2 | XP_013852587.1 | 1160 | 105 | ||
XM_021089841.1 | XP_020945500.1 | 1388 | 105 | |||
XM_021089842.1 | XP_020945501.1 | 847 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
