Assay Details
Gene Symbol: PROX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924492.3 | XP_001924527.1 | 1318 | 161 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
