Assay Details
Gene Symbol: HRH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078213.1 | XP_020933872.1 | 445 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
