Assay Details
Gene Symbol: AWAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021080857.1 | XP_020936516.1 | 1118 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
