Assay Details
Gene Symbol: ADGRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021085467.1 | XP_020941126.1 | 2012 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
