Assay Details
Gene Symbol: ALKAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087955.1 | XP_020943614.1 | 84 | 109 | ||
XM_021087956.1 | XP_020943615.1 | 84 | 109 | |||
XM_021087957.1 | XP_020943616.1 | 84 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021087955.1, XM_021087956.1, XM_021087957.1 transcripts.
Set Membership: |
![]() ![]() |
