Assay Details
Gene Symbol: ALKAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087955.1 | XP_020943614.1 | 522 | 61 | ||
XM_021087956.1 | XP_020943615.1 | 522 | 61 | |||
XM_021087957.1 | XP_020943616.1 | 522 | 61 | |||
XM_021087958.1 | XP_020943617.1 | 839 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
