Assay Details
Gene Symbol: TMPRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021091214.1 | XP_020946873.1 | 638 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
