Assay Details
Gene Symbol: PLA2G2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095473.1 | XP_020951132.1 | 69 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
