Assay Details
Gene Symbol: GFRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021098512.1 | XP_020954171.1 | 1338 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
