Assay Details
Gene Symbol: APOBEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126519.4 | XP_003126567.3 | 429 | 86 | ||
XM_013988403.2 | XP_013843857.1 | 432 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003126519.4, XM_013988403.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
