Assay Details
Gene Symbol: SPINK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129019.4 | XP_003129067.1 | 177 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
