Assay Details
Gene Symbol: GLRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003132832.6 | XP_003132880.3 | 3288 | 81 | ||
XM_013990304.2 | XP_013845758.1 | 1125 | 81 | |||
XM_021074179.1 | XP_020929838.1 | 2046 | 81 | |||
XM_021074180.1 | XP_020929839.1 | 2046 | 81 | |||
XM_021074181.1 | XP_020929840.1 | 1090 | 81 | |||
XM_021074182.1 | XP_020929841.1 | 3211 | 81 | |||
XM_021074183.1 | XP_020929842.1 | 2608 | 81 | |||
XM_021074184.1 | XP_020929843.1 | 1304 | 81 | |||
XM_021074186.1 | XP_020929845.1 | 1052 | 81 | |||
XM_021074187.1 | XP_020929846.1 | 2862 | 81 | |||
XM_021074188.1 | XP_020929847.1 | 2755 | 81 | |||
XM_021074189.1 | XP_020929848.1 | 1342 | 81 | |||
XM_021074190.1 | XP_020929849.1 | 1195 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
