Assay Details
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003357393.3 | XP_003357441.2 | 325 | 116 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
