Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003356327.4 | XP_003356375.2 | 651 | 108 | ||
XM_021096011.1 | XP_020951670.1 | 452 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021096011.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
