Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003356327.4 | XP_003356375.2 | 1135 | 125 | ||
XM_021096010.1 | XP_020951669.1 | 811 | 125 | |||
XM_021096011.1 | XP_020951670.1 | 799 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
