Assay Details
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001926554.5 | XP_001926589.2 | 1195 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
