Assay Details
Gene Symbol: MOB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096715.1 | XP_020952374.1 | 170 | 166 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021096715.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
