Assay Details
Gene Symbol: EIF2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003128668.4 | XP_003128716.2 | 262 | 62 | ||
XM_005656417.3 | XP_005656474.2 | 230 | 62 | |||
XM_021099531.1 | XP_020955190.1 | 230 | 62 | |||
GenBank mRNA | AK232859.1 | - | 1 - 2 | 242 | 62 | |
AY610188.1 | - | 1 - 2 | 219 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
