Assay Details
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021090517.1 | XP_020946176.1 | 1241 | 104 | ||
XM_021090518.1 | XP_020946177.1 | 1234 | 104 | |||
XM_021090519.1 | XP_020946178.1 | 1243 | 104 | |||
XM_021090520.1 | XP_020946179.1 | 1238 | 104 | |||
GenBank mRNA | AK232941.1 | - | 2 - 3 | 1001 | 104 | |
AK238486.1 | - | 2 - 3 | 1082 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
