Assay Details
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021090518.1 | XP_020946177.1 | 1468 | 96 | ||
XM_021090520.1 | XP_020946179.1 | 1443 | 96 | |||
XM_021090522.1 | XP_020946181.1 | 1359 | 96 | |||
GenBank mRNA | AK232941.1 | - | 4 - 5 | 1206 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
