Assay Details
Gene Symbol: AP1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086258.1 | XP_020941917.1 | 144 | 80 | ||
GenBank mRNA | AK232966.1 | - | 1 - 2 | 103 | 80 | |
AK348645.1 | - | 1 - 2 | 108 | 80 | ||
AK397060.1 | - | 1 - 2 | 118 | 80 | ||
AK400414.1 | - | 1 - 2 | 139 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086258.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
