Assay Details
Gene Symbol: CDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021084306.1 | XP_020939965.1 | 644 | 87 | ||
GenBank mRNA | AK233538.1 | - | 4 - 5 | 529 | 87 | |
AK391588.1 | - | 3 - 4 | 651 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
