Assay Details
Gene Symbol: ERF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127177.5 | XP_003127225.3 | 530 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
