Assay Details
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355494.5 | XP_003355542.1 | 148 | 83 | ||
XM_005663931.3 | XP_005663988.1 | 263 | 83 | |||
XM_021091883.1 | XP_020947542.1 | 266 | 83 | |||
XM_021091884.1 | XP_020947543.1 | 245 | 83 | |||
XM_021091885.1 | XP_020947544.1 | 245 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003355494.5, XM_005663931.3, XM_021091883.1, XM_021091884.1, XM_021091885.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
