Assay Details
Gene Symbol: H6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095286.1 | XP_020950945.1 | 340 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021095286.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
