Assay Details
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674572.3 | XP_005674629.2 | 216 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005674572.3 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
