Assay Details
Gene Symbol: GNA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086010.1 | XP_020941669.1 | 392 | 58 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
