Assay Details
Gene Symbol: GNA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086010.1 | XP_020941669.1 | 660 | 79 | ||
XM_021086013.1 | XP_020941672.1 | 993 | 79 | |||
XM_021086014.1 | XP_020941673.1 | 834 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
