Assay Details
Gene Symbol: NFE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003360190.4 | XP_003360238.2 | 1127 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
