Assay Details
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003361997.4 | XP_003362045.1 | 351 | 69 | ||
XM_021099316.1 | XP_020954975.1 | 586 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
