Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003359871.4 | XP_003359919.2 | 1749 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
