Assay Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977443.2 | XP_013832897.2 | 193 | 98 | ||
XM_013977445.2 | XP_013832899.2 | 132 | 98 | |||
XM_021099771.1 | XP_020955430.1 | 132 | 98 | |||
GenBank mRNA | AK233602.1 | - | 1 - 2 | 158 | 98 | |
AK239951.1 | - | 1 - 2 | 200 | 98 | ||
AK345197.1 | - | 2 - 3 | 235 | 98 | ||
AK348359.1 | - | 1 - 2 | 176 | 98 | ||
AK395015.1 | - | 1 - 2 | 161 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977443.2, XM_013977445.2, XM_021099771.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
