Assay Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977443.2 | XP_013832897.2 | 586 | 82 | ||
XM_013977446.2 | XP_013832900.2 | 411 | 82 | |||
XM_013977447.2 | XP_013832901.2 | 574 | 82 | |||
XM_013977449.2 | XP_013832903.2 | 2138 | 82 | |||
XM_021099771.1 | XP_020955430.1 | 630 | 82 | |||
XM_021099772.1 | XP_020955431.1 | 164 | 82 | |||
XM_021099774.1 | XP_020955433.1 | 8113 | 82 | |||
XM_021099775.1 | XP_020955434.1 | 1209 | 82 | |||
GenBank mRNA | AK233602.1 | - | 4 - 5 | 551 | 82 | |
AK239951.1 | - | 4 - 5 | 593 | 82 | ||
AK391349.1 | - | 1 - 2 | 276 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977443.2, XM_013977446.2, XM_013977447.2, XM_013977449.2, XM_021099771.1, XM_021099772.1, XM_021099774.1, XM_021099775.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
