Assay Details
Gene Symbol: SEPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064859.1 | XP_020920518.1 | 954 | 66 | ||
XM_021064860.1 | XP_020920519.1 | 1020 | 66 | |||
XM_021064861.1 | XP_020920520.1 | 1877 | 66 | |||
GenBank mRNA | AK234457.1 | - | 4 - 5 | 731 | 66 | |
AK345018.1 | - | 4 - 5 | 650 | 66 | ||
AY609937.1 | - | 5 - 6 | 682 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
