Assay Details
Gene Symbol: SEPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064859.1 | XP_020920518.1 | 1039 | 100 | ||
XM_021064860.1 | XP_020920519.1 | 1105 | 100 | |||
XM_021064861.1 | XP_020920520.1 | 1962 | 100 | |||
GenBank mRNA | AK234457.1 | - | 5 - 6 | 816 | 100 | |
AK345018.1 | - | 5 - 6 | 735 | 100 | ||
AY609937.1 | - | 6 - 7 | 767 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
