Assay Details
Gene Symbol: SEPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064859.1 | XP_020920518.1 | 1361 | 96 | ||
XM_021064860.1 | XP_020920519.1 | 1427 | 96 | |||
XM_021064861.1 | XP_020920520.1 | 2284 | 96 | |||
GenBank mRNA | AK234457.1 | - | 7 - 8 | 1138 | 96 | |
AY609937.1 | - | 8 - 9 | 1089 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
