Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK234511.1 | - | 1 - 2 | 198 | 94 | |
AK347653.1 | - | 1 - 2 | 186 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
