Assay Details
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124472.4 | XP_003124520.1 | 806 | 90 | ||
GenBank mRNA | AK234524.1 | - | 4 - 5 | 774 | 90 | |
AK400770.1 | - | 4 - 5 | 768 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
