Assay Details
Gene Symbol: DSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005659399.3 | XP_005659456.2 | 1149 | 80 | ||
XM_021091551.1 | XP_020947210.1 | 910 | 80 | |||
XM_021091554.1 | XP_020947213.1 | 1140 | 80 | |||
XM_021091561.1 | XP_020947220.1 | 372 | 80 | |||
XM_021091565.1 | XP_020947224.1 | 462 | 80 | |||
GenBank mRNA | AK234805.1 | - | 3 - 4 | 928 | 80 | |
AK348118.1 | - | 2 - 3 | 463 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
